Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B2778

Sigma-Aldrich

HBTU

Synonym(s):

N,N,N′,N′-Tetramethyl-O-(1H-benzotriazol-1-yl)uronium hexafluorophosphate, O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16F6N5OP
CAS Number:
Molecular Weight:
379.24
Beilstein/REAXYS Number:
7328329
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

200 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)C(\On1nnc2ccccc12)=[N+](/C)C

InChI

1S/C11H16N5O.F6P/c1-14(2)11(15(3)4)17-16-10-8-6-5-7-9(10)12-13-16;1-7(2,3,4,5)6/h5-8H,1-4H3;/q+1;-1

InChI key

UQYZFNUUOSSNKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Recent studies show that crystal as well as solution structure of this coupling agent is a guanidinium N-oxide and not an uronium compound; Coupling reagent for peptide synthesis; advantages are: very low racemization, simple reaction conditions, very short reaction time, and high yields.

Other Notes

This product has been replaced by 12804-ALDRICH | HBTU ≥98.0% (T)

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1A

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service