Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B0253

Sigma-Aldrich

Brucine

Synonym(s):

10,11-Dimethoxystrychnine, 2,3-Dimethoxystrychnidin-10-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N2O4
CAS Number:
Molecular Weight:
394.46
Beilstein/REAXYS Number:
63046
EC Number:
MDL number:
UNSPSC Code:
12352000

mp

175-178 °C (dec.) (lit.)

SMILES string

[H][C@@]12CC(=O)N3c4cc(OC)c(OC)cc4[C@]56CC[N@H]7CC(=CCO1)[C@]([H])(C[C@@H]57)[C@]2([H])[C@]36[H]

InChI

1S/C23H26N2O4/c1-27-16-8-14-15(9-17(16)28-2)25-20(26)10-18-21-13-7-19-23(14,22(21)25)4-5-24(19)11-12(13)3-6-29-18/h3,8-9,13,18-19,21-22H,4-7,10-11H2,1-2H3/t13-,18-,19-,21-,22-,23+/m0/s1

Inchi Key

RRKTZKIUPZVBMF-IBTVXLQLSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Aquatic Chronic 3

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service