Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A9035

Sigma-Aldrich

Atriopeptin II rat

Synonym(s):

Atrial Natriuretic Peptide Fragment 5-27], Atriopeptin-23, rat, rANF (103-125)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C98H156N34O32S2
CAS Number:
Molecular Weight:
2386.63
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)CNC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC(=O)CNC1=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](N)CO)[C@@H](C)CC

InChI

1S/C98H156N34O32S2/c1-8-48(5)76-93(161)115-38-71(140)116-50(7)78(146)120-56(26-27-68(100)137)83(151)128-63(42-134)82(150)114-39-73(142)118-58(31-47(3)4)80(148)113-40-74(143)119-66(91(159)125-61(34-69(101)138)87(155)129-65(44-136)89(157)124-60(33-52-21-14-11-15-22-52)86(154)122-57(95(163)164)25-18-30-110-98(106)107)45-165-166-46-67(130-90(158)64(43-135)127-79(147)53(99)41-133)92(160)123-59(32-51-19-12-10-13-20-51)81(149)112-36-70(139)111-37-72(141)117-54(23-16-28-108-96(102)103)84(152)132-77(49(6)9-2)94(162)126-62(35-75(144)145)88(156)121-55(85(153)131-76)24-17-29-109-97(104)105/h10-15,19-22,47-50,53-67,76-77,133-136H,8-9,16-18,23-46,99H2,1-7H3,(H2,100,137)(H2,101,138)(H,111,139)(H,112,149)(H,113,148)(H,114,150)(H,115,161)(H,116,140)(H,117,141)(H,118,142)(H,119,143)(H,120,146)(H,121,156)(H,122,154)(H,123,160)(H,124,157)(H,125,159)(H,126,162)(H,127,147)(H,128,151)(H,129,155)(H,130,158)(H,131,153)(H,132,152)(H,144,145)(H,163,164)(H4,102,103,108)(H4,104,105,109)(H4,106,107,110)/t48-,49-,50-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,76-,77-/m0/s1

InChI key

MAGGONBKRAPXMU-KJZYAOAHSA-N

Gene Information

rat ... Nppa(24602)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg

Biochem/physiol Actions

Activates particulate guanylyl cyclase; cardiac atrial peptide which relaxes both intestinal and vascular smooth muscle.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M V Ovchinnikov et al.
Bioorganicheskaia khimiia, 14(6), 777-782 (1988-06-01)
A series of atrial natriuretic peptides, viz., alpha-r-ANF, APII, APIII, and des-Ser5, Ser6-APII, have been obtained by condensation of earlier synthesized fragments promoted with complex F. Acetylaminomethyl protecting group were removed from cystein residues with simultaneous cyclization of the peptide.
J C Millar et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 13(1), 1-11 (1997-02-01)
The effects of atriopeptin (AP) on intraocular pressure (IOP), aqueous humor formation (AHF) and ciliary cyclic GMP in bovine eyes perfused in vitro through a ciliary artery were investigated. AHF was monitored fluorophotometrically by perfusing the anterior chamber with Bárány's
P A Whitson et al.
Journal of cellular physiology, 146(1), 43-51 (1991-01-01)
Atrial natriuretic peptide (ANP) binding and ANP-induced increases in cyclic guanosine monophosphate (cGMP) levels have been observed in brain microvessels (Chabrier et al., 1987; Steardo and Nathanson, 1987), suggesting that this fluid-regulating hormone may play a role in the fluid
L B Fernandes et al.
European journal of pharmacology, 212(2-3), 187-194 (1992-03-03)
Three peptide components of atrial natriuretic factor (ANF) caused relaxation of carbachol-contracted guinea-pig isolated tracheal smooth muscle. These were the 1-28, 5-28 and 5-27 peptide sequences (ANF(1-28), ANF-(5-28) and ANF-(5-27)). The peptides were 10-30 times more potent in epithelium-denuded than
M G Currie et al.
Science (New York, N.Y.), 223(4631), 67-69 (1984-01-06)
Mammalian cardiac atria have several biologically active peptides that exert profound effects on sodium excretion, urine volume, and smooth muscle tone. In the present study two such peptides of low molecular weight were purified and separated from each other on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service