A7560
iso-A-Pentasaccharide
from human urine, ≥90%
Synonym(s):
α-GalNAc-(1→3)-(α-Fuc-[1→2])-β-Gal-(1→3)-(α-Fuc-[1→4])-Glc, A-Leb-Pentasaccharide
About This Item
biological source
human urine
assay
≥90%
storage temp.
2-8°C
SMILES string
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](CO)OC(O)[C@H](O)[C@H]2O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4NC(C)=O)[C@H]3O[C@@H]5O[C@@H](C)[C@@H](O)[C@@H](O)[C@@H]5O)[C@@H](O)[C@H](O)[C@@H]1O
InChI
1S/C32H55NO24/c1-7-14(38)19(43)21(45)30(49-7)54-24-12(6-36)51-28(48)23(47)26(24)56-32-27(57-31-22(46)20(44)15(39)8(2)50-31)25(17(41)11(5-35)53-32)55-29-13(33-9(3)37)18(42)16(40)10(4-34)52-29/h7-8,10-32,34-36,38-48H,4-6H2,1-3H3,(H,33,37)/t7-,8-,10+,11+,12+,13+,14+,15+,16-,17-,18+,19+,20+,21-,22-,23+,24+,25-,26+,27+,28?,29+,30-,31-,32-/m0/s1
InChI key
ZBXMDMBUJYNWCZ-NLMCABFBSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service