Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7181

Sigma-Aldrich

Adrenocorticotropic Hormone Tyr-Fragment 4-9 human, rat

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H65N13O11S
CAS Number:
Molecular Weight:
1068.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(N)Cc1ccc(O)cc1)C(=O)NC(CCC(O)=O)C(=O)NC(Cc2cnc[nH]2)C(=O)NC(Cc3ccccc3)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc4c[nH]c5ccccc45)C(O)=O

Gene Information

human ... POMC(5443)

Amino Acid Sequence

Tyr-Met-Glu-His-Phe-Arg-Trp

Biochem/physiol Actions

Analog of ACTH (4-9) that can be iodinated for receptor and histochemical studies

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service