Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4766

Sigma-Aldrich

Acetic acid-d

Synonym(s):

mono-Deuteroacetic acid, Acetic (acid-d), Monodeuteroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOD
CAS Number:
Molecular Weight:
61.06
Beilstein/REAXYS Number:
1739249
EC Number:
MDL number:
UNSPSC Code:
12352200

isotopic purity

98 atom % D

mass shift

M+1

SMILES string

[2H]OC(C)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i/hD

InChI key

QTBSBXVTEAMEQO-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

104.0 °F - closed cup

flash_point_c

40 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service