Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4405

Sigma-Aldrich

Anisole

Synonym(s):

Methoxybenzene, Methyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H5
CAS Number:
Molecular Weight:
108.14
Beilstein/REAXYS Number:
506892
EC Number:
MDL number:
UNSPSC Code:
12352209

vapor density

3.7 (vs air)

vapor pressure

10 mmHg ( 42.2 °C)

autoignition temp.

887 °F

refractive index

n20/D 1.516 (lit.)

bp

154 °C (lit.)

mp

−37 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1

InChI

1S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

RDOXTESZEPMUJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Kohji Omata et al.
Journal of combinatorial chemistry, 12(4), 435-444 (2010-05-22)
To study the synergistic effect on the acidity and activity of Si-Al-Zr ternary oxide system, the mixture design of experiments was applied to prepare the ternary oxides by a sol-gel method, and rapid screening on dehydration of isopropanol and Friedel-Crafts
Xiaohui Jin et al.
Water research, 46(19), 6519-6530 (2012-10-20)
Second-order reaction rate constants of micropollutants with ozone (k(O3)) and hydroxyl radicals (k(OH)) are essential for evaluating their removal efficiencies from water during ozonation and advanced oxidation processes. Kinetic data are unavailable for many of the emerging micropollutants. Twenty-four micropollutants
Laszlo Hollosi et al.
Journal of chromatography. A, 1218(1), 23-31 (2010-12-03)
European food legislation defines a set of 16 polycyclic aromatic hydrocarbons (PAHs) as of high concern for human health. The EU set contains structurally very similar PAHs with ring numbers between 4 and 6, and so raises some separation aspects
Adam F Lee et al.
Dalton transactions (Cambridge, England : 2003), 39(43), 10473-10482 (2010-10-05)
Size-controlled, catalytically active PVP-stabilised Pd nanoparticles have been studied by operando liquid phase XAS during the Suzuki cross-coupling of iodonanisole and phenylboronic acid in MeOH-toluene using KOMe base. XAS reveals nanoparticles are stable to metal leaching throughout the reaction, with
Shouhui Zhang et al.
The Journal of organic chemistry, 75(19), 6732-6735 (2010-09-09)
A CuI-catalyzed direct access to sulfides from disulfides via C-H bond cleavage of di- or trimethoxybenzene is described. The procedure utilizes O(2) as a clean and cheap oxidant. Direct selenation of the C-H bond also took place under this procedure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service