Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3921

Sigma-Aldrich

Arachidonamide

10 mg/mL in ethanol, ~97%, low-melting solid

Synonym(s):

5,8,11,14-Eicosatetraenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H33NO
CAS Number:
Molecular Weight:
303.48
MDL number:
UNSPSC Code:
12352200

assay

~97%

form

low-melting solid

concentration

10 mg/mL in ethanol

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(N)=O

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corey, E.J., et al.
Journal of the American Chemical Society, 106, 1503-1503 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service