Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A3477

Sigma-Aldrich

AH 11110

>98% (HPLC), solid

Synonym(s):

1-[Biphenyl-2-yloxy]-4-imino-4-piperidin-1-yl-butan-2-ol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O2 · HCl
CAS Number:
Molecular Weight:
374.90
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

>98% (HPLC)

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

slightly yellow

solubility

H2O: soluble ~2.5 mg/mL
DMSO: insoluble

application(s)

forensics and toxicology
veterinary

originator

GlaxoSmithKline

storage temp.

−20°C

SMILES string

Cl[H].OC(COc1ccccc1-c2ccccc2)CC(=N)N3CCCCC3

InChI

1S/C21H26N2O2.ClH/c22-21(23-13-7-2-8-14-23)15-18(24)16-25-20-12-6-5-11-19(20)17-9-3-1-4-10-17;/h1,3-6,9-12,18,22,24H,2,7-8,13-16H2;1H

InChI key

WWHGFIXWBLHHQB-UHFFFAOYSA-N

Biochem/physiol Actions

α1B-adrenoceptor ligand

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Zhang et al.
Circulation research, 89(9), 815-822 (2001-10-27)
In vivo studies have suggested that norepinephrine (NE) directly contributes to normal vascular wall growth and worsening of hypertrophy, atherosclerosis, and restenosis. However, it is unknown whether these effects are secondary to hemodynamic changes caused by systemic NE or alpha-adrenoceptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service