Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3180

Sigma-Aldrich

Amyloid β-Protein Fragment 12-28

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C89H135N25O25
CAS Number:
Molecular Weight:
1955.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(CCCCN)NC(=O)C(CCC(N)=O)NC(=O)C(Cc1cnc[nH]1)NC(=O)C(Cc2cnc[nH]2)NC(=O)C(N)C(C)C)C(=O)NC(C(C)C)C(=O)NC(Cc3ccccc3)C(=O)NC(Cc4ccccc4)C(=O)NC(C)C(=O)NC(CCC(O)=O)C(=O)NC(CC(O)=O)C(=O)NC(C(C)C)C(=O)NCC(=O)NC(CO)C(=O)NC(CC(N)=O)C(=O)NC(CCCCN)C(O)=O

Gene Information

human ... APP(351)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys

Biochem/physiol Actions

Impairs post-training memory in mice upon injection into limbic system structures.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Castaño et al.
Biochemical and biophysical research communications, 141(2), 782-789 (1986-12-15)
Two synthetic peptides corresponding to the reported 28-residue sequence of Alzheimer's Disease beta-protein (SP28) and to residues 12-28 (SP17) were used to form fibrils in vitro. Synthetic fibrils bound Congo Red and closely resembled amyloid fibrils isolated from leptomeninges and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service