Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2513

Sigma-Aldrich

N-Acetyl-L-tyrosine

>99% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
223.23
Beilstein/REAXYS Number:
2697172
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

>99% (TLC)

mp

149-152 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C11H13NO4/c1-7(13)12-10(11(15)16)6-8-2-4-9(14)5-3-8/h2-5,10,14H,6H2,1H3,(H,12,13)(H,15,16)/t10-/m0/s1

InChI key

CAHKINHBCWCHCF-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-acetyl tyrosyluria caused by parenteral or enteral administration of N-acetyl-L-tyrosine: differentiation from hereditary and acquired tyrosinemias.
Stanley H Korman et al.
Journal of pediatric gastroenterology and nutrition, 39(1), 95-100 (2004-06-10)
W Wang et al.
The Biochemical journal, 340 ( Pt 1), 329-336 (1999-05-07)
The formation of a reversible adsorption complex between a dimer of N-acetyl-L-tyrosine [di-(N-acetyl-L-tyrosine), (NAT)2] and horseradish peroxidase (HRP) compound II (CII) was demonstrated using a kinetic approach. A specific KIIm value (0.58 mM) was deduced for this step from stopped-flow
L John Hoffer et al.
JPEN. Journal of parenteral and enteral nutrition, 27(6), 419-422 (2003-11-19)
N-acetyl-L-tyrosine (NAT) is commonly used in place of tyrosine in parenteral nutrition, but human studies carried out to date indicate considerable amounts of it are excreted unchanged in the urine. NAT retention has not been well studied in parenterally fed
[Recording the delta-pH-generating biochemical reactions by light-addressed sensors with Ta2O5 dielectrics].
A N Reshetilov et al.
Doklady Akademii nauk, 342(5), 700-702 (1995-06-01)
Rakesh Kumar et al.
European journal of medicinal chemistry, 42(4), 503-510 (2006-12-26)
The development of type 2 diabetes in obese individuals is linked to lipid accumulation in non-adipose tissues. A series of N-acetyl-L-tyrosine derivatives were synthesized and evaluated for PPAR transactivation. Compounds 4d and 4f were found to show better PPARalpha transactivation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service