All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H24N7O12P
CAS Number:
Molecular Weight:
573.41
MDL number:
UNSPSC Code:
12352204
storage temp.
−20°C
SMILES string
N.Nc1ncnc2n(cnc12)C3OC(CO)C(O)C3OP(O)(=O)OCC4OC(C(O)C4O)N5C=CC(=O)NC5=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service