Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A185

Sigma-Aldrich

Ambenonium dichloride

solid

Synonym(s):

N,N′-[(1,2-Dioxo-1,2-ethanediyl)bis(imino-2,1-ethanediyl)] bis[2-chloro-N,N-diethylbenzene-methanaminium]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H42Cl4N4O2
CAS Number:
Molecular Weight:
608.47
EC Number:
MDL number:
UNSPSC Code:
12352200

form

solid

color

white

solubility

H2O: soluble

SMILES string

[Cl-].[Cl-].CC[N+](CC)(CCNC(=O)C(=O)NCC[N+](CC)(CC)Cc1ccccc1Cl)Cc2ccccc2Cl


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Hodge et al.
Molecular pharmacology, 41(5), 937-942 (1992-05-01)
Steady state patterns of inhibition of purified human erythrocyte acetylcholinesterase by three inhibitors were analyzed. Edrophonium acted essentially as a competitive inhibitor, whereas tacrine and ambenonium gave mixed competitive and uncompetitive inhibition with acetylthiocholine as substrate. Inhibition constants for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service