Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0539

Sigma-Aldrich

(Aminomethyl)phosphonic acid

≥99%

Synonym(s):

AMPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2P(O)(OH)2
CAS Number:
Molecular Weight:
111.04
Beilstein/REAXYS Number:
1701219
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥99%

SMILES string

NCP(O)(O)=O

InChI

1S/CH6NO3P/c2-1-6(3,4)5/h1-2H2,(H2,3,4,5)

InChI key

MGRVRXRGTBOSHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Analog of glycine; degradation product of glyphosate.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emmanuel O Mogusu et al.
Analytical and bioanalytical chemistry, 407(18), 5249-5260 (2015-05-15)
To assess sources and degradation of the herbicide glyphosate [N-(phosphonomethyl) glycine] and its metabolite AMPA (aminomethylphosphonic acid), concentration measurements are often inconclusive and even (13)C/(12)C analysis alone may give limited information. To advance isotope ratio analysis of an additional element

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service