Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96632

Sigma-Aldrich

Bis(2,2′-bipyridine)-4,4′-dicarboxybipyridine-ruthenium di(N-succinimidyl ester) bis(hexafluorophosphate)

BioReagent, suitable for fluorescence

Synonym(s):

Ru(bpy)2(dcbpy-di O-Su-ester)(PF6)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H30F12N8O8P2Ru
CAS Number:
Molecular Weight:
1141.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

fluorescence

λex 458 nm; λem 647 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

shipped in

dry ice

Storage temp.

−20°C

SMILES string

[Ru++].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.c1ccc(nc1)-c2ccccn2.c3ccc(nc3)-c4ccccn4.O=C5CCC(=O)N5OC(=O)c6ccnc(c6)-c7cc(ccn7)C(=O)ON8C(=O)CCC8=O

InChI

1S/C20H14N4O8.2C10H8N2.2F6P.Ru/c25-15-1-2-16(26)23(15)31-19(29)11-5-7-21-13(9-11)14-10-12(6-8-22-14)20(30)32-24-17(27)3-4-18(24)28;2*1-3-7-11-9(5-1)10-6-2-4-8-12-10;2*1-7(2,3,4,5)6;/h5-10H,1-4H2;2*1-8H;;;/q;;;2*-1;+2

InChI key

HIPJTYPALXAMBX-UHFFFAOYSA-N

Application

Ruthenium-ligand belonging to a new class of long-lived fluorophores for protein hydrodynamics.

Features and Benefits

Sigma-Aldrich offers a series of reactive dyes based on ruthenium complexes for use in fluorescence polarization assays and fluorescence lifetime imaging applications. They have been designed specifically to have long fluorescence lifetimes. These dyes were first synthesized in the labs of Dr. Joseph Lakowicz at the Center for Fluorescence Microscopy. These laser-excitable metal ligand complexes have fluorescence lifetimes near 500 nanoseconds. These dyes can be used to label amines on biomolecules under mild conditions. Advantages of these dyes as protein labels include high photostability, good water solubility, a lack of dye-dye interactions and large Stokes′ shifts. In addition, the fluorescence signal of long-lived fluorophores can be gated to eliminate the emission from short-lifetime fluorophores and autofluorescence from cells and biomolecules to further improve sensitivity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Terpetschnig et al.
Biophysical journal, 68(1), 342-350 (1995-01-01)
We describe the use of asymmetric Ru-ligand complexes as a new class of luminescent probes that can be used to measure rotational motions of proteins. These complexes are known to display luminescent lifetimes ranging from 10 to 4000 ns. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service