Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92320

Sigma-Aldrich

Ethyl-β-D-thiogalactopyranoside

≥98.0% (HPLC)

Synonym(s):

Ethyl β-D-thiogalactoside, Ethyl-1-thio-β-D-Gal, Ethyl-1-thio-β-D-galactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O5S
CAS Number:
Molecular Weight:
224.27
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D -24.0±2.0, c = 1% (w/v) in water

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

CCS[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C8H16O5S/c1-2-14-8-7(12)6(11)5(10)4(3-9)13-8/h4-12H,2-3H2,1H3/t4-,5+,6+,7-,8+/m1/s1

InChI key

CHAHFVCHPSPXOE-HNEXDWKRSA-N

Application

Ethyl-β-D-thiogalactopyranoside is a derivitized thiogalactopyranoside structurally related to the gene expression inducer isopropyl-1-thio-β-D-galactopyranoside (IPTG).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fu'ad T Haddadin et al.
Biotechnology and bioengineering, 90(2), 127-153 (2005-03-03)
The transcriptome profiles for wild-type (plasmid-free) and recombinant (plasmid-bearing) Escherichia coli during well-controlled synchronized high-cell-density fed-batch cultures were analyzed by DNA microarrays. It was observed that the growth phase significantly affected the transcriptome profiles, and the transcriptome profiles were significantly
H Sheng et al.
Cancer research, 61(6), 2670-2675 (2001-04-06)
Cyclooxygenase (COX) 2 expression is regulated via the Ras signaling pathway, and induction of mutated Ras rapidly increases COX-2 levels in intestinal epithelial cells. Protein kinase B (Akt/PKB) is an important effector of Ras signaling and a critical component of
Teruo Tanaka et al.
Journal of bacteriology, 187(13), 4315-4326 (2005-06-22)
Plasmid pL32 from the Natto strain of Bacillus subtilis belongs to a group of low-copy-number plasmids in gram-positive bacteria that replicate via a theta mechanism of replication. We studied the DNA region encoding the replication protein, RepN, of pLS32, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service