Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82860

Sigma-Aldrich

Pyridoxal hydrochloride

≥99.5% (AT)

Synonym(s):

3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxaldehyde hydrochloride, PL HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO3 · HCl
CAS Number:
Molecular Weight:
203.62
Beilstein/REAXYS Number:
3656994
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥99.5% (AT)

loss

≤0.6% loss on drying

mp

173 °C (dec.) (lit.)

anion traces

sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤250 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤15 mg/kg
Cu: ≤5 mg/kg
Fe: ≤15 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

Cl[H].[H]C(=O)c1c(CO)cnc(C)c1O

InChI

1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,4,10,12H,3H2,1H3;1H

InChI key

FCHXJFJNDJXENQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For the labeling of amino acids and their detection in picomolar amounts; Coenzymes and Cofactors, vol. 1: Vitamin B6, Pyridoxal Phosphate

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service