Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72540

Sigma-Aldrich

Nitr 5 tetrasodium salt

≥95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H27N3Na4O15
CAS Number:
Molecular Weight:
773.52
Beilstein/REAXYS Number:
6953888
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥95% (TLC)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1ccc(N(CC([O-])=O)CC([O-])=O)c(OCCOc2cc(ccc2N(CC([O-])=O)CC([O-])=O)C(O)c3cc4OCOc4cc3[N+]([O-])=O)c1

Other Notes

Water-soluble photosensitive calcium chelator that releases calcium on photolysis[1][2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.R. Adams et al.
Journal of the American Chemical Society, 110, 3212-3212 (1988)
A M Gurney et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(10), 3496-3500 (1987-05-01)
Although Ca2+ is a well-established intracellular messenger, there are many questions concerning the kinetics and spatial localization of its effects. Such problems may now be approached with the photosensitive Ca2+ chelator nitr-5. The Ca2+ affinity of this molecule decreases by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service