Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63951

Sigma-Aldrich

4,4′-[(8,16-Dihydro-8,16-dioxodibenzo[a,j]perylene-2,10-diyl)dioxy]dibutyric acid

BioReagent, suitable for fluorescence, ≥90% (CH)

Synonym(s):

2,10-Bis(3-carboxypropyloxy)dibenzo[a,j]perylene-8,16-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H26O8
CAS Number:
Molecular Weight:
586.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

assay

≥90% (CH)

mp

231-235 °C

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

OC(=O)CCCOc1ccc-2c(c1)C(=O)c3cccc4c5-c6ccc(OCCCC(O)=O)cc6C(=O)c7cccc(c-2c34)c57

InChI

1S/C36H26O8/c37-29(38)9-3-15-43-19-12-14-22-27(17-19)35(41)25-7-1-5-23-31-21-13-11-20(44-16-4-10-30(39)40)18-28(21)36(42)26-8-2-6-24(34(26)31)32(22)33(23)25/h1-2,5-8,11-14,17-18H,3-4,9-10,15-16H2,(H,37,38)(H,39,40)

InChI key

XZWIRUDAKQRKFW-UHFFFAOYSA-N

Analysis Note

luminescence: Emax 668 nm (Eex: 620 nm in chloroform)

Other Notes

Highly photostable, NIR fluorescent label with larger Stokes shift than is usual for cyanine dyes and which is maximally excited, (max CHCl3) 620 nm, by He-Ne laser (630 nm) or diode laser (635 nm).[1]

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.W. Deady et al.
Synthetic Communications, 30, 803-803 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service