Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63565

Sigma-Aldrich

D-Mannitol

≥98.0% (sum of enantiomers, HPLC)

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein/REAXYS Number:
1721898
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

assay

≥98.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D +24±1° in borax solution acc. to Ph Eur

mp

165-167 °C
167-170 °C (lit.)

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian-Rong Li et al.
European journal of medicinal chemistry, 43(8), 1688-1695 (2007-11-27)
Two novel rare earth complexes, Y(III) complex (1) and Eu(III) complex (2), with naringenin-2-hydroxy benzoyl hydrazone ligand were synthesized and characterized. The interaction of the two metal complexes and the free ligand with calf thymus DNA (CT DNA) was investigated
Markus Bollinger et al.
Journal of medicinal chemistry, 55(2), 871-882 (2011-12-22)
Intervention in integrin-mediated cell adhesion and integrin signaling pathways is an ongoing area of research in medicinal chemistry and drug development. One key element in integrin-ligand interaction is the coordination of the bivalent cation at the metal ion-dependent adhesion site
Christopher J Burns et al.
Bioorganic & medicinal chemistry letters, 19(20), 5887-5892 (2009-09-19)
A series of phenylaminopyrimidines has been identified as inhibitors of Janus kinases (JAKs). Development of this initial series led to the potent JAK2/JAK1 inhibitor CYT387 (N-(cyanomethyl)-4-[2-[[4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]-benzamide). Details of synthesis and SAR studies of these compounds are reported.
Jayanta Chatterjee et al.
Journal of medicinal chemistry, 50(24), 5878-5881 (2007-11-02)
An unselective cyclic peptide integrin ligand was sequentially N-methylated by a designed approach, where only the externally oriented (solvent exposed) amide bonds were N-methylated. The N-methylation resulted in tremendous enhancement in selectivity among the different integrin receptor subtypes (alpha5beta1, alphavbeta3
Bao-Ning Su et al.
Journal of natural products, 68(4), 592-595 (2005-04-23)
Purification of a n-BuOH-soluble partition of the MeOH extract of Morinda citrifolia (Noni) fruits led to the isolation of two new iridoid glucosides, 6alpha-hydroxyadoxoside (1) and 6beta,7beta-epoxy-8-epi-splendoside (2), as well as 17 known compounds, americanin A (3), narcissoside (4), asperuloside

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service