Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59900C

SAFC

Water - Cell Culture Grade

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein/REAXYS Number:
2050024
EC Number:
MDL number:
UNSPSC Code:
12352207

description

for research or for further manufacturing use
meets criteria for WFI, endotoxin < 0.25 EU/mL

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets all USP and EP criteria for Water for Injection (WFI). It has undergone purification via activated carbon, reverse osmosis, deionization, ultraviolet disinfection, and distillation. For use in the preparation of cell culture media, and cell suspension and washing solutions.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siyu Wang et al.
Ecotoxicology and environmental safety, 120, 193-197 (2015-06-19)
A total of 120 children (58 males and 62 females) between the ages of 2 and 17 years were randomly selected from Wuhan City and Wufeng County in Hubei Province, China. We gathered hand SD samples from these children using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service