Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59205C

HEPES Buffer Solution (1M)

Synonym(s):

HEPES solution, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

description

for research or for further manufacturing use

form

liquid

useful pH range

6.8-8.2

storage temp.

2-8°C

SMILES string

OCCN1CCN(CCS(=O)(O)=O)CC1

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

HEPES is used in many media because it has more buffering capacity in the physiologic pH range of 7.2 - 7.4 at 37 C than sodium bicarbonate. It is commonly added at concentrations between 10 mM and 25 mM. Higher levels cause cytotoxicity. Because bicarbonate is nutritionally necessary for most cells, HEPES should be added in addition to, not in place of, sodium bicarbonate.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Marwitz et al.
Laboratory investigation; a journal of technical methods and pathology, 94(8), 927-933 (2014-06-17)
Alterations in the DNA methylome are characteristic for numerous diseases and a typical hallmark of cancer. Therefore, DNA methylation is currently under investigation in research labs and has also entered diagnostics. Recently, protocols like the BeadChip technology have become commercially

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service