Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57569

Sigma-Aldrich

myo-Inositol

BioUltra, ≥99.5% (HPLC)

Synonym(s):

1,2,3,4,5,6-Hexahydroxycyclohexane, i-Inositol, meso-Inositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1907329
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

vapor density

6.2 (vs air)

product line

BioUltra

assay

≥99.5% (HPLC)

impurities

insoluble matter, passes filter test

ign. residue

≤0.2%

loss

≤0.5% loss on drying, 20 °C (HV)

pH

5.0-7.0 (25 °C, 0.5 M in H2O)

mp

222-227 °C (lit.)
224-227 °C

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.1
λ: 280 nm Amax: 0.1

SMILES string

O[C@H]1[C@H](O)[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4+,5-,6-

InChI key

CDAISMWEOUEBRE-GPIVLXJGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Myo-Inositol, an organic osmolyte, is used to improve ovarian function in women with polycystic ovary syndrome (PCOS). Myo-Inositol may be used to study the H+-myo-inositol transporter SLC2A13, a Potential Marker for Cancer Stem Cells in an Oral Squamous Cell Carcinoma.

Biochem/physiol Actions

A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers.

Other Notes

Metabolism and function of myo-inositol and inositol phospholipids[1]; Review [2][3]; Component of the liquid stationary phase in the GLC separation of nitroxylene and xylenol isomers[4]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ss
Inositol & Phosphoinositides: Metab.& Reg., eds.J.E. Bleasdale et al. (1985)
B J Holub
Annual review of nutrition, 6, 563-597 (1986-01-01)
Alterations in the level of dietary inositol can significantly influence the concentration of free inositol and inositol-containing phospholipid in the circulation and in selected mammalian tissues and cells. The 1-stearoyl 2-arachidonyl molecular species that commonly predominates in cellular phosphoinositides may
Inositol & Phosphoinositides: Metab.& Reg., eds.J.E. Bleasdale et al. (1985)
Separation behaviour of some isomeric organic compounds on sugars, sugar alcohols and their mixed phases by gas-liquid chromatography.
Ono, A.
Journal of Chromatography A, 197, 251-251 (1980)
M Utriainen et al.
Journal of neuro-oncology, 62(3), 329-338 (2003-06-05)
The signal of choline containing compounds (Cho) in proton magnetic resonance spectroscopy (1H-MRS) is elevated in brain tumors. [11C]choline uptake as assessed using positron emission tomography (PET) has also been suggested to be higher in brain tumors than in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service