Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55377

Sigma-Aldrich

Octaethyleneglycol monododecyl ether 1 mM solution

Synonym(s):

Octaethyleneglycol monododecyl ether solution, Detergent Screening Solution 22/Kit-No 66317

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

non-ionic

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryuichi Ueoka et al.
Bioorganic & medicinal chemistry letters, 18(16), 4578-4580 (2008-08-02)
A good correlation between fifty-percent inhibitory concentration (IC(50)) of hybrid liposomes (HL) composed of dimyristoylphosphatidylcholine and polyoxyethylene(n) dodecyl ether on the growth of MOLT-4/IIIB cells (MOLT-4 cells chronically infected with human immunodeficiency virus (HIV)) and the membrane fluidity of HL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service