Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53704

Sigma-Aldrich

Calcium chloride 0.1 M solution

Synonym(s):

Calcium chloride solution, Additive Screening Solution 21/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

Cl[Ca]Cl

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

Inchi Key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Maheswari Jayakannan et al.
Journal of experimental botany, 66(7), 1865-1875 (2015-01-24)
The role of endogenous salicylic acid (SA) signalling cascades in plant responses to salt and oxidative stresses is unclear. Arabidopsis SA signalling mutants, namely npr1-5 (non-expresser of pathogenesis related gene1), which lacks NPR1-dependent SA signalling, and nudt7 (nudix hydrolase7), which
J Philippe et al.
International journal of obesity (2005), 39(2), 295-302 (2014-06-04)
A significant proportion of severe familial forms of obesity remain genetically elusive. Taking advantage of our unique cohort of multigenerational obese families, we aimed to assess the contribution of rare mutations in 29 common obesity-associated genes to familial obesity, and
Mohsen Akbari et al.
Advanced functional materials, 24(26), 4060-4067 (2014-11-21)
The fabrication of cell-laden structures with anisotropic mechanical properties while having a precise control over the distribution of different cell types within the constructs is important for many tissue engineering applications. Automated textile technologies for making fabrics allow simultaneous control
Michael A Goren et al.
Nature communications, 5, 5115-5115 (2014-10-09)
Opsin, the rhodopsin apoprotein, was recently shown to be an ATP-independent flippase (or scramblase) that equilibrates phospholipids across photoreceptor disc membranes in mammalian retina, a process required for disc homoeostasis. Here we show that scrambling is a constitutive activity of
S Zhen et al.
Gene therapy, 22(5), 404-412 (2015-02-06)
The current therapies to treat hepatitis B virus (HBV) infection are limited. Recently, clustered regularly interspaced short palindromic repeat (CRISPR) systems, originally identified in bacteria and archaea, have been found to consist of an RNA-based adaptive immune system that degrades

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service