Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51783

Sigma-Aldrich

3,5-Diiodo-L-tyrosine dihydrate

≥98.0% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9I2NO3 · 2H2O
CAS Number:
Molecular Weight:
469.01
Beilstein/REAXYS Number:
2218691
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

≥98.0% (NT)

optical activity

[α]/D +1.2±0.4°, c = 5% in 1 M HCl

mp

199-201 °C (dec.)
200 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

N[C@@H](Cc1cc(I)c(O)c(I)c1)C(O)=O

InChI

1S/C9H9I2NO3/c10-5-1-4(2-6(11)8(5)13)3-7(12)9(14)15/h1-2,7,13H,3,12H2,(H,14,15)/t7-/m0/s1

InChI key

NYPYHUZRZVSYKL-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intermediate in the biosynthesis and alternative pathways of metabolism of thyroid hormones. Substrate for the assay of halogenated tyrosine and thyroid hormone aminotransferase

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Nakano
Methods in Enzymology, 17A, 660-660 (1970)
Purification and properties of halogenated tyrosine and thyroid hormone transaminase from rat kidney mitochondria.
M Nakano
The Journal of biological chemistry, 242(1), 73-81 (1967-01-10)
A Balsam et al.
The Journal of clinical investigation, 72(4), 1234-1245 (1983-10-01)
Studies were performed to elucidate the nature of the pathway of hepatic thyroxine (T4) metabolism that is activated by inhibitors of liver catalase. For this purpose, the metabolism of T4 in homogenates of rat liver was monitored with T4 labeled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service