Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49746

Sigma-Aldrich

Anthrose

≥98.0% (TLC)

Synonym(s):

4,6-Dideoxy-4-(3-hydroxy-3-methylbutanamido)-2-O-methyl-D-glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO6
CAS Number:
Molecular Weight:
277.31
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

assay

≥98.0% (TLC)

form

solid

storage temp.

−20°C

SMILES string

C[C@H]1OC(O)[C@H](O)[C@@H](O)[C@@H]1NC(=O)CC(C)(C)O

InChI

1S/C11H21NO6/c1-5-7(8(14)9(15)10(16)18-5)12-6(13)4-11(2,3)17/h5,7-10,14-17H,4H2,1-3H3,(H,12,13)/t5-,7-,8+,9-,10?/m1/s1

InChI key

UUVASJJCLBEXCO-QNRYFBKSSA-N

Application

Anthrose, a nonreducing dideoxy-glucopyranose, has recently been identified as a unique component of the bacillus anthracis exosporium and Bacillus cereus endospores. Anthrose is used in anthrax research.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rina Saksena et al.
Carbohydrate research, 340(9), 1591-1600 (2005-06-01)
The key step in the first chemical synthesis of anthrose (16) and its methyl alpha- (6) and beta-glycoside (22) was inversion of configuration at C-2 in triflates 10, 2, and 18, respectively, obtained from the common intermediate, methyl 4-azido-3-O-benzyl-4,6-dideoxy-alpha-D-mannopyranoside (1).
Rachel L Kubiak et al.
Biochemistry, 51(4), 867-878 (2012-01-10)
The unusual dideoxy sugar d-anthrose has been identified as an important component in the endospores of infectious agents such as Bacillus anthracis and Bacillus cereus. Specifically, it is the terminal sugar on the bacterium's exosporium, and it provides a point
James M Daubenspeck et al.
The Journal of biological chemistry, 279(30), 30945-30953 (2004-05-21)
Spores of Bacillus anthracis, the causative agent of anthrax, are enclosed by a prominent loose fitting layer called the exosporium. The exosporium consists of a basal layer and an external hairlike nap. The filaments of the nap are composed of
John G Bruno et al.
Journal of fluorescence, 22(3), 915-924 (2012-01-06)
A library of 92 DNA aptamer sequences was developed against Bacillus anthracis (nonpathogenic Sterne strain) spores and anthrose sugar immobilized on magnetic beads. The selected DNA sequences were studied for similarities and potential binding pockets between the B. anthracis spore
Michael H Norris et al.
PLoS biology, 18(12), e3001052-e3001052 (2020-12-29)
Bacillus anthracis, a spore-forming gram-positive bacterium, causes anthrax. The external surface of the exosporium is coated with glycosylated proteins. The sugar additions are capped with the unique monosaccharide anthrose. The West African Group (WAG) B. anthracis have mutations rendering them

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service