Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44211

Sigma-Aldrich

Nitrilotriacetic acid tri(N-succinimidyl) ester

purum, trifunctional, amine-reactive cross-linking agent, ≥97.0% (N)

Synonym(s):

Tri(N-succinimidyl) nitrilotriacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N4O12
CAS Number:
Molecular Weight:
482.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

assay

≥97.0% (N)

mp

~169 °C (dec.)

storage temp.

−20°C

SMILES string

O=C(CN(CC(=O)ON1C(=O)CCC1=O)CC(=O)ON2C(=O)CCC2=O)ON3C(=O)CCC3=O

InChI

1S/C18H18N4O12/c23-10-1-2-11(24)20(10)32-16(29)7-19(8-17(30)33-21-12(25)3-4-13(21)26)9-18(31)34-22-14(27)5-6-15(22)28/h1-9H2

InChI key

CYIJYHIEQLQITN-UHFFFAOYSA-N

Application

Crosslinking agent for compounds carrying an amino group

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service