Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43568

Sigma-Aldrich

Cesium chloride 1 M solution

Synonym(s):

Cesium chloride solution, Additive Screening Solution 13/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

density

1.13 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[Cs]

InChI

1S/Cs

InChI key

TVFDJXOCXUVLDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piotr Jurkiewicz et al.
Biochimica et biophysica acta, 1818(3), 609-616 (2011-12-14)
Effects of alkali metal chlorides on the properties of mixed negatively charged lipid bilayers are experimentally measured and numerically simulated. Addition of 20mol% of negatively charged phosphatidylserine to zwitterionic phosphatidylcholine strengthens adsorption of monovalent cations revealing their specificity, in the
Aleksander W Ballo et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(15), 5106-5119 (2012-04-13)
We studied how conduction delays of action potentials in an unmyelinated axon depended on the history of activity and how this dependence was changed by the neuromodulator dopamine (DA). The pyloric dilator axons of the stomatogastric nervous system in the
Sandra Aedo et al.
Antimicrobial agents and chemotherapy, 56(11), 5458-5464 (2012-08-08)
DNA topoisomerases are important targets in anticancer and antibacterial therapy because drugs can initiate cell death by stabilizing the transient covalent topoisomerase-DNA complex. In this study, we employed a method that uses CsCl density gradient centrifugation to separate unbound from
Philip A Gurnev et al.
Langmuir : the ACS journal of surfaces and colloids, 28(45), 15824-15830 (2012-10-24)
We demonstrate that the cation-selective channel formed by gramicidin A can be used as a reliable sensor for studying the multivalent ion accumulation at the surfaces of charged lipid membranes and the "charge inversion" phenomenon. In asymmetrically charged membranes with
Cambrian Y Liu et al.
Journal of neurophysiology, 108(5), 1318-1334 (2012-06-01)
Mammals rely on their acute olfactory sense for their survival. The most anterior olfactory subsystem in the nose, the Grueneberg ganglion (GG), plays a role in detecting alarm pheromone, cold, and urinary compounds. GG neurons respond homogeneously to these stimuli

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service