Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42897

Sigma-Aldrich

Isopropyl 1-thio-β-D-glucuronide sodium salt

≥97.0% (TLC)

Synonym(s):

IPTGlcA · Na, Isopropyl β-D-thioglucuronic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NaO6S
CAS Number:
Molecular Weight:
274.27
UNSPSC Code:
12352201
PubChem Substance ID:

assay

≥97.0% (TLC)

form

powder

optical activity

[α]/D -86.0±2.5°, c = 1 in H2O

storage temp.

−20°C

SMILES string

CC(C)SC1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O[Na]

InChI

1S/C9H16O6S.Na/c1-3(2)16-9-6(12)4(10)5(11)7(15-9)8(13)14;/h3-7,9-12H,1-2H3,(H,13,14);/q;+1/p-1/t4-,5-,6+,7-,9?;/m0./s1

InChI key

KBUAMHFDNNUCET-JYYOLFTLSA-M

Application

Isopropyl β-D-thioglucuronic acid (Isopropyl 1-thio-β-D-glucuronide) is closely related to the lac operon inducer, isopropyl 1-thio-β-D-glalactoside. Isopropyl β-D-thioglucuronic acid is used to induce β-D-glucosidase(s) for detection of bacteriophage via chromogenic assays.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service