Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42387

Sigma-Aldrich

Triethyleneglycol monooctyl ether 1 mM solution

Synonym(s):

Triethylene glycol monooctyl ether solution, Detergent Screening Solution 09/Kit-No 66317

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service