41261
N,N-Dimethyloctylamine N-oxide 100 mM solution
Synonym(s):
N,N-Dimethyloctylamine N-oxide solution, Detergent Screening Solution 40/Kit-No 66317
About This Item
storage temp.
2-8°C
SMILES string
CCCCCCCC[N+](C)(C)[O-]
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service