40913
Nonaethyleneglycol monododecyl ether 1 mM solution
Synonym(s):
Nonaethyleneglycol monododecyl ether solution, Detergent Screening Solution 23/Kit-No 66317
About This Item
description
non-ionic
storage temp.
2-8°C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service