Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31903

Sigma-Aldrich

N,N-Diethylpropargylamine

for protein sequence analysis, ≥99.0% (GC)

Synonym(s):

3-Diethylamino-1-propyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2N(CH2CH3)2
CAS Number:
Molecular Weight:
111.18
Beilstein/REAXYS Number:
1098468
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for protein sequence analysis

assay

≥99.0% (GC)

bp

118-120 °C (lit.)

density

0.80 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN(CC)CC#C

InChI

1S/C7H13N/c1-4-7-8(5-2)6-3/h1H,5-7H2,2-3H3

InChI key

JZJXKEWVUBVOEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

68.0 °F - closed cup

flash_point_c

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Préaux et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 360(11), 1595-1604 (1979-11-01)
The isolation of beta-lactoglobulin from milk of the goat is described. The purified protein was checked for purity and has been characterized by its gross composition and end groups. The native or the modified protein was then degraded by tryptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service