25517
Sodium tartrate dibasic dihydrate
meets analytical specification of E335, ≥99%
Synonym(s):
L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate
About This Item
assay
≥99%
quality
meets analytical specification of E335
impurities
residual solvents, in accordance
≤0.001% ammonium (NH4)
loss
≤17% loss on drying, 150 °C, 4 h
pH
7.0-7.5 (20 °C, 1%)
anion traces
chloride (Cl-): ≤50 mg/kg
oxalate (as oxalic acid): ≤100 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
As: ≤2 mg/kg
Ca: ≤100 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
K: ≤200 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg
SMILES string
[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
InChI
1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1
InChI key
FGJLAJMGHXGFDE-DGFHWNFOSA-L
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service