Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18472

Sigma-Aldrich

Betaine solution

0.1 M

Synonym(s):

Additive Screening Solution 40/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2COO
CAS Number:
Molecular Weight:
117.15
Beilstein/REAXYS Number:
3537113
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

concentration

0.1 M

density

1.00 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

C[N+](C)(C)CC([O-])=O

InChI

1S/C5H11NO2/c1-6(2,3)4-5(7)8/h4H2,1-3H3

InChI key

KWIUHFFTVRNATP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernd C Schwahn et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 17(3), 512-514 (2003-01-29)
Hyperhomocysteinemia, a proposed risk factor for cardiovascular disease, is also observed in other common disorders. The most frequent genetic cause of hyperhomocysteinemia is a mutated methylenetetrahydrofolate reductase (MTHFR), predominantly when folate status is impaired. MTHFR synthesizes a major methyl donor
Pål I Holm et al.
Clinical chemistry, 49(2), 286-294 (2003-02-01)
The quaternary ammonium compounds, choline and betaine, and dimethylglycine (DMG) reside along a metabolic pathway linked to the synthesis of neurotransmitters and membrane phospholipids and to homocysteine remethylation and, therefore, folate status. Lack of a convenient, high-throughput method for the
Per Magne Ueland
Journal of inherited metabolic disease, 34(1), 3-15 (2010-05-07)
Choline is an essential nutrient, but is also formed by de novo synthesis. Choline and its derivatives serve as components of structural lipoproteins, blood and membrane lipids, and as a precursor of the neurotransmitter acetylcholine. Pre-and postnatal choline availability is
Per Magne Ueland et al.
Clinical chemistry and laboratory medicine, 43(10), 1069-1075 (2005-10-04)
Betaine serves as a methyl donor in a reaction converting homocysteine to methionine, catalysed by the enzyme betaine-homocysteine methyltransferase. It has been used for years to lower the concentration of plasma total homocysteine (tHcy) in patients with homocystinuria, and has
M R Olthof et al.
Current drug metabolism, 6(1), 15-22 (2005-02-22)
High plasma concentrations of homocysteine may increase risk of cardiovascular disease. Folic acid lowers plasma homocysteine by 25% maximally, because 5-methyltetrahydrofolate is a methyl donor in the remethylation of homocysteine to methionine. Betaine (trimethylglycine) is also a methyl donor in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service