Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16459

Supelco

N,N′-Bis(1-hexylheptyl)-perylene-3,4:9,10-bis-(dicarboximide)

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H62N2O4
CAS Number:
Molecular Weight:
755.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98.0% (TLC)

form

solid

fluorescence

λex 523 nm; λem 535 nm in ethanol

storage temp.

2-8°C

SMILES string

CCCCCCC(CCCCCC)N1C(=O)c2ccc3c4ccc5C(=O)N(C(CCCCCC)CCCCCC)C(=O)c6ccc(c7ccc(C1=O)c2c37)c4c56

InChI

1S/C50H62N2O4/c1-5-9-13-17-21-33(22-18-14-10-6-2)51-47(53)39-29-25-35-37-27-31-41-46-42(32-28-38(44(37)46)36-26-30-40(48(51)54)45(39)43(35)36)50(56)52(49(41)55)34(23-19-15-11-7-3)24-20-16-12-8-4/h25-34H,5-24H2,1-4H3

InChI key

NJEZHCHNQGICLU-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service