Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2254

Sigma-Aldrich

1,1′-Dimethyl-4,4′-bipyridinium dichloride hydrate

Synonym(s):

Gramoxone, Methyl viologen dichloride, Paraquat dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14Cl2N2 · xH2O
CAS Number:
Molecular Weight:
257.16 (anhydrous basis)
Beilstein/REAXYS Number:
3752782
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

SMILES string

C[N+]1=CC=C(C2=CC=[N+](C)C=C2)C=C1.[Cl-].[Cl-].O

InChI

1S/C12H14N2.2ClH.H2O/c1-13-7-3-11(4-8-13)12-5-9-14(2)10-6-12;;;/h3-10H,1-2H3;2*1H;1H2/q+2;;;/p-2

InChI key

RLGVBTFWTZJYIL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Lungs, Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service