Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3570

Sigma-Aldrich

TFLZn

≥90% (TLC)

Synonym(s):

4-[([6-Methoxy-2-methyl-8-quinolinyl]amino)sulfonyl]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500

assay

≥90% (TLC)

color

off-white to pink

solubility

aqueous base: soluble

storage temp.

−20°C

SMILES string

COc1cc(NS(=O)(=O)c2ccc(cc2)C(O)=O)c3nc(C)ccc3c1

Biochem/physiol Actions

May be used to measure intracellular Zn++ levels

Analysis Note

The TFLZn-Zn complex is fluorescent, with an excitation at 380 nm, and emission at 510 nm

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service