Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S6628

Supelco

Silica gel

high-purity grade, Type H, 10-40 μm, suitable for thin layer chromatography (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:

grade

high-purity grade

type

Type H

contains

Al2O3 as binder

technique(s)

thin layer chromatography (TLC): suitable

particle size

10-40 μm

bp

2230 °C

mp

>1600 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service