P6758
Potassium phthalate monobasic
≥99%
Synonym(s):
KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate
About This Item
Recommended Products
assay
≥99%
pH
4
mp
295-300 °C (dec.) (lit.)
SMILES string
[K+].OC(=O)c1ccccc1C([O-])=O
InChI
1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1
InChI key
IWZKICVEHNUQTL-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Other Notes
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
387.1 °F - Pensky-Martens closed cup
flash_point_c
197.3 °C - Pensky-Martens closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service