Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N2880

Sigma-Aldrich

Neutral Red

practical grade

Synonym(s):

3-Amino-7-dimethylamino-2-methylphenazine hydrochloride, Basic Red 5, Toluylene red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78
Colour Index Number:
50040
Beilstein/REAXYS Number:
3918740
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

practical grade

composition

Dye content, ~60%

technique(s)

titration: suitable

pH

6.8-8.0, red to yellow

mp

290 °C (dec.) (lit.)

SMILES string

Cl.CN(C)c1ccc2nc3cc(C)c(N)cc3nc2c1

InChI

1S/C15H16N4.ClH/c1-9-6-13-15(8-11(9)16)18-14-7-10(19(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H

InChI key

PGSADBUBUOPOJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful as an indicator for preparing neutral red paper, and as a biological stain.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina T Siirin et al.
The American journal of tropical medicine and hygiene, 79(6), 955-962 (2008-12-05)
The efficacy of a recombinant subunit West Nile (WN) vaccine candidate was determined in a hamster model of encephalitis. Animals included young, aged, and immunocompromised animals in an effort to simulate key groups at risk of WN virus-induced disease. Groups
Adrivanio Baranoski et al.
Toxicology mechanisms and methods, 25(9), 672-679 (2015-05-15)
The β-glucans (β-G) are polysaccharides produced by various organisms, and sulfation of β-G renders them more soluble. With the objective to assess the effects of sulfated and non-sulfated β-G extracted from Agaricus brasiliensis in MCF-7 cells, assays were used to
Timothy D Harrington et al.
Bioresource technology, 192, 689-695 (2015-06-22)
The aim of this work was to elucidate the mechanism of mediated microbial electrosynthesis via neutral red from an electrode to fermenting Escherichia coli cultures in a bioelectrochemical system. Chemical reduction of NAD(+) by reduced neutral red did not occur
Mary F Fontana et al.
PLoS pathogens, 7(2), e1001289-e1001289 (2011-03-11)
The intracellular bacterial pathogen Legionella pneumophila causes an inflammatory pneumonia called Legionnaires' Disease. For virulence, L. pneumophila requires a Dot/Icm type IV secretion system that translocates bacterial effectors to the host cytosol. L. pneumophila lacking the Dot/Icm system is recognized
Sonia Soloneski et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(5), 834-844 (2015-03-31)
In vitro effects of the carbamates pirimicarb and zineb and their formulations Aficida® (50% pirimicarb) and Azzurro® (70% zineb), respectively, were evaluated in Chinese hamster ovary (CHO-K1) cells. Whereas the cytokinesis-blocked micronucleus cytome assay was employed to test for genotoxicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service