Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8006

Sigma-Aldrich

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate

Synonym(s):

MBTH hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N3S · HCl · xH2O
CAS Number:
Molecular Weight:
215.70 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100

mp

276-278 °C (dec.) (lit.)

Storage temp.

2-8°C

SMILES string

Cl[H].[H]O[H].CN1\C(Sc2ccccc12)=N\N

InChI

1S/C8H9N3S.ClH.H2O/c1-11-6-4-2-3-5-7(6)12-8(11)10-9;;/h2-5H,9H2,1H3;1H;1H2/b10-8-;;

Inchi Key

IYXXQOGEFHAQGU-GPWRMYTLSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service