Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2629

Sigma-Aldrich

3-O-Methylfluorescein phosphate cyclohexylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15O8P · C6H13N
CAS Number:
Molecular Weight:
525.49
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

storage temp.

−20°C

SMILES string

NC1CCCCC1.COc2ccc3c(Oc4cc(OP(O)(O)=O)ccc4C35OC(=O)c6ccccc56)c2

InChI

1S/C21H15O8P.C6H13N/c1-26-12-6-8-16-18(10-12)27-19-11-13(29-30(23,24)25)7-9-17(19)21(16)15-5-3-2-4-14(15)20(22)28-21;7-6-4-2-1-3-5-6/h2-11H,1H3,(H2,23,24,25);6H,1-5,7H2

InChI key

SJPHZMRXIHOGOW-UHFFFAOYSA-N

Application

For use in the fluorimetric determination of phosphatase.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An automated fluorometric assay for alkaline phosphatase using 3-O-methylfluorescein phosphate.
H D Hill et al.
Analytical biochemistry, 24(1), 9-17 (1968-07-01)
Lok Hang Mak et al.
Methods (San Diego, Calif.), 77-78, 63-68 (2015-03-10)
PTEN (phosphatase and tensin homologue deleted on chromosome 10) is well known as a tumour suppressor. It's PI(3,4,5)P3 lipid phosphatase activity is an important counteracting mechanism in PI 3-kinase (phosphoinositide 3-kinase) signalling. Furthermore, PTEN lies upstream of Akt kinase, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service