Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L7161

Sigma-Aldrich

Lime

meets USP testing specifications

Synonym(s):

Calcium oxide, Lime, Quicklime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaO
CAS Number:
Molecular Weight:
56.08
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

agency

meets USP testing specifications

bp

2850 °C (lit.)

density

3.3 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

O=[Ca]

InChI

1S/Ca.O

Inchi Key

ODINCKMPIJJUCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Nazaret González-Alcaraz et al.
Journal of environmental management, 116, 72-80 (2013-01-08)
The aim of this study was to identify the effectiveness of liming in combination with vegetation for the recovery of slightly acidic, saline soils of eutrophic wetlands affected by mine wastes, under fluctuating flooding conditions. Simulated soil profiles were constructed
Sebastião Roberto Soares et al.
Waste management (New York, N.Y.), 33(1), 175-183 (2012-11-06)
The establishment of rules to manage Health Care Waste (HCW) is a challenge for the public sector. Regulatory agencies must ensure the safety of waste management alternatives for two very different profiles of generators: (1) hospitals, which concentrate the production
Vasilije Manovic et al.
International journal of environmental research and public health, 7(8), 3129-3140 (2010-10-16)
This paper presents a review of the research on CO(2) capture by lime-based looping cycles undertaken at CanmetENERGY's (Ottawa, Canada) research laboratories. This is a new and very promising technology that may help in mitigation of global warming and climate
Kunlun Zhang et al.
Journal of hazardous materials, 243, 278-285 (2012-11-20)
Tetrabromobisphenol A (TBBPA) is the most widely used brominated flame retardant (BFR), which has received more and more concerns due to its high lipophilicity, persistency and endocrine disrupting property in the environment. Considering the possible need for the safe disposal
Yong-Lu Meng et al.
Bioresource technology, 128, 305-309 (2012-12-01)
A solid Ca/Al composite oxide-based alkaline catalyst containing Ca(12)Al(14)O(33) and CaO was prepared by chemical synthesis and thermal activation from sodium aluminate solution and calcium hydroxide emulsion. The effect of calcination temperatures ranging from 120 °C to 1000 °C on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service