About This Item
Recommended Products
agency
meets USP testing specifications
vapor density
9 (vs air)
vapor pressure
0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)
assay
99.8-100.5%
resistivity
1.3E15 μΩ-cm
impurities
≤0.05% Nonvolatile residue
bp
184 °C (lit.)
mp
113 °C (lit.)
anion traces
chloride, bromide (as Cl)-: ≤0.028%
application(s)
pharmaceutical (small molecule)
SMILES string
II
InChI
1S/I2/c1-2
InChI key
PNDPGZBMCMUPRI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
target_organs
Respiratory system, Thyroid
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service