Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I3147

Supelco

Chromoionophore I

suitable for function test

Synonym(s):

3-Octadecanoylimino-7-(diethylamino)-1,2-benzophenoxazine, 9-(Diethylamino)-5-(octadecanoylimino)-5H-benzo[a]phenoxazine, N-Octadecanoyl-Nile blue, ETH 5294

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H53N3O2
CAS Number:
Molecular Weight:
583.85
Beilstein/REAXYS Number:
3576427
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

mp

91-93 °C (lit.)

fluorescence

λex 614 nm; λem 663 nm (protonated)

suitability

suitable for function test

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)\N=C1/C=C2Oc3cc(ccc3N=C2c4ccccc14)N(CC)CC

InChI

1S/C38H53N3O2/c1-4-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25-37(42)39-34-29-36-38(32-24-22-21-23-31(32)34)40-33-27-26-30(28-35(33)43-36)41(5-2)6-3/h21-24,26-29H,4-20,25H2,1-3H3/b39-34+

InChI key

MYWBZQJIXVRCJC-JQJRJOTLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Assay of K+ and of Ca2+ activity with solvent polymeric optode membrane based on ETH 5294 and potassium ionophore I (valinomycin)

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service