Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G7043

Sigma-Aldrich

Glycerin

meets EP, BP, JP, USP testing specifications, synthetic

Synonym(s):

Glycerol, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein/REAXYS Number:
635685
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

biological source

synthetic

agency

meets BP testing specifications
meets EP testing specifications
meets JP testing specifications
meets USP testing specifications

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

autoignition temp.

698 °F

quality

meets EP, BP, JP, USP testing specifications

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

390.2 °F - Pensky-Martens closed cup

flash_point_c

199 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad S Eram et al.
Biochimica et biophysica acta, 1850(9), 1842-1848 (2015-05-24)
Dysregulation of methylation of lysine 36 on histone H3 (H3K36) have been implicated in a variety of diseases including cancers. ASH1L and SETD2 are two enzymes among others that catalyze H3K36 methylation. H3K4 methylation has also been reported for ASH1L.
Benoît Ranchoux et al.
The American journal of pathology, 185(2), 356-371 (2014-12-17)
Pulmonary veno-occlusive disease (PVOD) is an uncommon form of pulmonary hypertension (PH) characterized by progressive obstruction of small pulmonary veins and a dismal prognosis. Limited case series have reported a possible association between different chemotherapeutic agents and PVOD. We evaluated
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target
Tian Qiu et al.
Nature communications, 5, 5119-5119 (2014-11-05)
Biological microorganisms swim with flagella and cilia that execute nonreciprocal motions for low Reynolds number (Re) propulsion in viscous fluids. This symmetry requirement is a consequence of Purcell's scallop theorem, which complicates the actuation scheme needed by microswimmers. However, most
T W Chew et al.
Oncogene, 33(21), 2717-2727 (2013-07-03)
RAS and Rho small GTPases are key molecular switches that control cell dynamics, cell growth and tissue development through their distinct signaling pathways. Although much has been learnt about their individual functions in both cell and animal models, the physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service