D61563
1,2-Dichloroethane
ReagentPlus®, 99%
Synonym(s):
Ethylene chloride, Ethylene dichloride
About This Item
Recommended Products
vapor density
3.4 (20 °C, vs air)
vapor pressure
87 mmHg ( 25 °C)
product line
ReagentPlus®
assay
99%
autoignition temp.
775 °F
expl. lim.
15.6 %
refractive index
n20/D 1.444 (lit.)
bp
83 °C (lit.)
mp
−35 °C (lit.)
density
1.256 g/mL at 25 °C (lit.)
SMILES string
ClCCCl
InChI
1S/C2H4Cl2/c3-1-2-4/h1-2H2
InChI key
WSLDOOZREJYCGB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
55.4 °F - closed cup
flash_point_c
13 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service