Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D0681

Sigma-Aldrich

Diethanolamine

BioXtra, ≥98.5%

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein/REAXYS Number:
605315
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

product line

BioXtra

assay

≥98.5%

form

liquid

autoignition temp.

689 °F

expl. lim.

10.6 %

impurities

≤0.0005% Phosphorus (P)

ign. residue

≤0.005%

refractive index

n20/D 1.477 (lit.)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

1.097 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

target_organs

Kidney,Liver,Blood

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

280.4 °F - closed cup

flash_point_c

138 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Conway et al.
Environmental science & technology, 46(13), 7422-7429 (2012-05-25)
The kinetics of the fast reversible carbamate formation reaction of CO(2)(aq) with a series of substituted cyclic secondary amines as well as the noncyclic secondary amine diethanolamine (DEA) has been investigated using the stopped-flow spectrophotometric technique at 25.0 °C. The
Siou-Yin Chen et al.
Inorganic chemistry, 51(8), 4448-4457 (2012-04-05)
The synthesis, X-ray crystallography, magnetic properties, and high-field electron paramagnetic resonance (HFEPR) of a new heptanuclear manganese complex [Mn(7)(heamp)(6)](ClO(4))(2)·4CH(2)Cl(2)·H(2)O (complex 2), in which heampH(3) is 2-[N,N-di(2-hydroxyethyl)aminomethyl]phenol (compound 1), is reported. Complex 2 has a hexagonal, disk-shaped topology and contains six
Honghui Yu et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 2902-2910 (2014-08-15)
Anti-D can prevent immunization to the RhD Ag on RBCs, a phenomenon commonly termed Ab-mediated immune suppression (AMIS). The most accepted theory to explain this effect has been the rapid clearance of RBCs. In mouse models using SRBC, these xenogeneic
Valentyna V Semenaka et al.
Inorganic chemistry, 49(12), 5460-5471 (2010-06-15)
Two new tetranuclear complexes, [Zn(2)Cr(2)(NCS)(4)(Dea)(2)(HDea)(2)].4DMSO (1; DMSO = dimethyl sulfoxide) and [Zn(2)Cr(2)(NCS)(4)(Dea)(2)(HDea)(2)].2CH(3)CN (2), were prepared from zinc oxide, Reinecke's salt, NH(4)[Cr(NCS)(4)(NH(3))(2)].H(2)O, ammonium thiocyanate, and a nonaqueous solution of diethanolamine (H(2)Dea) in a reaction carried out under open air. Both compounds
Bjørn Henrik Hansen et al.
Aquatic toxicology (Amsterdam, Netherlands), 99(2), 212-222 (2010-06-12)
Alkanolamines are surface-active chemicals used in a wide range of industrial, agricultural and pharmaceutical applications and products. Of particular interest is the use of alkanolamines such as diethanolamine (DEA) in the removal of CO(2) from natural gas and for CO(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service